Learn More
Melford Cefoperazone
Description
Cefoperazone Sodium Salt is a third-generation cephalosporin antibiotic. Cefoperazone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).
Cefoperazone is effective against a broad range of bacteria, both Gram-positive and Gram-negative. Cefoperazone Sodium Salt is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion allows for the specific growth of bacteria lacking resistance to cefoperazone.
Spécification
Spécification
| Nom chimique ou matériau | Cefoperazone |
| CAS | 62893-20-3 |
| Quantité | 10 g |
| Formule moléculaire | C25H26N9NaO8S2 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.