missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine dihydrochloride, MedChemExpress

Product Code. 16421836
Encompass_Preferred
Click to view available options
Quantité:
100 mg
200 mg
500 mg
This item is not returnable. View return policy

Product Code. 16421836

Brand: Medchem Express HY17042A

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

MedChemExpress Cetirizine dihydrochloride, a second-generation antihistamine and the carboxylated metabolite of hydroxyzine, is a specific, orally active and long-acting histamine H1-receptor antagonist. Cetirizine dihydrochloride marks antiallergic properties and inhibits eosinophil chemotaxis during the allergic response.

Cetirizine dihydrochloride, a second-generation antihistamine and the carboxylated metabolite of hydroxyzine, is a specific, orally active and long-acting histamine H1-receptor antagonist. Cetirizine dihydrochloride marks antiallergic properties and inhibits eosinophil chemotaxis during the allergic response.

Specifications

Nom chimique ou matériau Cetirizine dihydrochloride
Forme physique Solid
CAS 83881-52-1
Couleur Blanc
Quantité 500 mg
Notes de qualité de la pureté Research
Formule moléculaire C21H27Cl3N2O3
À utiliser avec (application) COVID-19-immunoregulation
Synonyme P071
Informations sur la solubilité DMSO : 100 mg/mL (216.54 mM; Need ultrasonic) ∣H2O : 100 mg/mL (216.54 mM; Need ultrasonic)
SMILES O=C(O)COCCN1CCN(C(C2=CC=C(Cl)C=C2)C3=CC=CC=C3)CC1.Cl.Cl
Poids moléculaire (g/mol) 461.81
Poids de la formule 461.81
Pourcentage de pureté 99.17%
Qualité Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.