Learn More
Sampling Systems™ Chemical Spoon and Spatula
Code produit. 17942112 Tous les produits Sampling Systems Produits
Click to view available options
Longueur (métrique):
154 mm
201 mm
280 mm
317 mm
Conditionnement:
1 pièce
Code produit. 17942112
Marque: Sampling Systems™ A625150
Il en reste null
Ajouter au panier
Ajouter au panier
This versatile chemical spoon and spatula has a robust design and is made from 304 stainless steel. It can be used as both a spoon and a spatula to allow for greater versatility in the sampling of both powder and liquid products.
  • No sharp edges or crevices, making it ideal for use in food, pharma and chemical companies
  • Available in a range of sizes
  • Heavy duty design for comfortable use
  • Ideal for measuring, sampling and dosing powders and liquids
Spécification
Stérilité Non stérile
Autoclavable Autoclavable
Jetable Reusable
Type de produit Spoon
Matériau Acier inoxydable
Longueur (métrique) 154 mm