Learn More
DiscoverX ChemiScreen™ mGlu2 Metabotropic Glutamate Receptor Stable Cell Line
Click to view available options
Quantité:
2 flacons
Conditionnement:
2 ampoules
Description
The ChemiScreen™ mGlu2 Metabotropic Glutamate Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. glutamate) based activation of GRM2 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GRM2 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of GRM2 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.
Spécification
Spécification
| Entrée du dosage | Calcium Flux |
| Lignée cellulaire | Chem-1 |
| À utiliser avec (application) | Cell Lines |
| Agoniste primaire | L-Glutamate |
| Quantité | 2 flacons |
| Entrée cible | GRM2 |
| Numéro d’adhésion | NM_000839 |