Learn More
BOLA Chromatography Adapters
Click to view available options
Diamètre du filetage:
18
25
32
45
Diamètre (métrique) externe:
1,6 mm
3,2 mm
Conditionnement:
1 pièce
Description
- Black screw cap made of PPS with GL thread.
- Body made of PTFE with one port with female thread UNF ¼ in. 28G for connection of Mini Fittings.
- A possible unevenness of the bottle neck is adjusted by an o-ring behind an elastic sealing lip, and the bottle is closed tightly.
- The product is only exposed to the body of the adaptor.
- Very good chemical resistance, for working temperatures up to max. +200°C.
Spécification
Spécification
| Diamètre (métrique) interne | 0.8 mm |
| Diamètre du filetage | 18 |
| Diamètre (métrique) externe | 1,6 mm |
| Autoclavable | Autoclavable |