Learn More
Cole-Parmer Cleanroom Large-Bore Female Luer to Hose Barb Fittings, Straight Adapter, White Nylon
Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,188 po
0,25 po
0,313 po
Conditionnement:
10 pièces
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Spécification
Spécification
| Certifications/Conformité | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diamètre (impériale) interne | 0,313 po |
| Matériau | Nylon |
| Quantité d’emballage | 10/Pk. |
| Admission | Large Bore Female Luer, Hose Barb |
| Couleur | blanc |