Learn More
Cole-Parmer Cleanroom Large-Bore Female Luer Plugs, Straight Adapter
Tous les produits Cole-Parmer Produits
Click to view available options
Matériau:
CrystalVu (CVU)
Kynar
Nylon
Polypropylène
Couleur:
Naturel
Transparent
blanc
Conditionnement:
10 pièces
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers
Spécification
Spécification
| Certifications/Conformité | Non-DEHP;Non-Phthalate;RoHS;USP |
| Matériau | CrystalVu (CVU) |
| Quantité d’emballage | 10/Pk. |
| Admission | Large Bore Female Luer, Plug |
| Couleur | Transparent |