Learn More
Cole-Parmer Cleanroom Port Disk Hose Barb Fittings, Flat Plate, Natural Kynar™ Flex
Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,125 po
0,188 po
0,25 po
0,313 po
0,5 po
0,625 po
0,75 po
1 po
3/8 in.
5/32 po
Conditionnement:
10 pièces
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Flexible, smooth flat base for easy attachment. Ultrasonic, radio frequency, or thermal weld to Kynar bags. Meets FDA requirements 21 CFR 177.1520(c), 21 CFR 176.170(c), 21 CFR 177.2600(c)(4)(i), NSF/ANSI Standard 51 and 61, and RoHS compliant
Spécification
Spécification
| Certifications/Conformité | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diamètre (impériale) interne | 0,75 po |
| Quantité d’emballage | 10/Pk. |
| Port de sortie | Port Disk x Hose Barb |
| Méthode de stérilisation | 121°C |
| Forme | Structuré |
| Couleur | Naturel |