Learn More
STEMCELL Technologies ClonaCell™-HY Liquid HAT Selection Medium
Code produit. 17148211
Encompass
Click to view available options
Quantité:
500 ml
Conditionnement:
500mL
Code produit. 17148211
Marque: STEMCELL Technologies 03831
Hybridoma selection medium with HAT (serum-containing)
  • ClonaCell™-HY Liquid HAT Selection Medium is a serum-containing medium with hypoxanthine, aminopterin, and thymidine (HAT) for the selection of hybridomas following the fusion of lymphocytes and myeloma cells
  • This medium has been verified for use in mouse and rat hydridoma development and reportedly is compatible for production and cloning of myelomas and/or hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster
Spécification
Type de cellule Hybridomes
À utiliser avec (application) Cell Culture; Hybridoma Generation
Quantité 500 ml