missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobicistat, MedChemExpress
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Description
Cobicistat is a potent and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC50s of 30-285 nM. Cobicistat is a pharmacokinetic enhancer which increases the overall absorption of several HIV medications.
Specifications
Specifications
| Nom chimique ou matériau | Cobicistat |
| Forme physique | Solid |
| CAS | 1004316-88-4 |
| Couleur | Jaune clair |
| Quantité | 100 mg |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C40H53N7O5S2 |
| À utiliser avec (application) | COVID-19-anti-virus |
| Synonyme | GS-9350 |
| Informations sur la solubilité | DMSO : 250 mg/mL (322.16 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction