Learn More
BOLA Cordless Pumps for Acids and Caustic Solutions
Click to view available options
Longueur totale (métrique):
400 mm
600 mm
Conditionnement:
1 pièce
Description
- Made of polypropylene, PTFE, Hastelloy™, driven by two commercial 1,5 V batteries (we recommend the use of rechargeable batteries).
- Powerful – pumping capacity of up to 6 litres per minute free flowing
- Compact construction
- Battery operated and therefore usable anywhere
- Easy handling
- Very light weight (only 500 g including batteries)
- Low-risk pumping
- Also suitable for narrow mouth vessels with ground joint 29/32 or thread GL 45, carboys or barrels
Spécification
Spécification
| Applications | For pumping low viscous liquids |
| Taille de l’ouverture (métrique) | 25 mm |
| Longueur totale (métrique) | 400 mm |
| Matériau | Polypropylene |