Learn More
STEMCELL Technologies CoV Protease Substrate-1 EDANS
Click to view available options
Quantité:
100 tests
1 000 tests
Conditionnement:
100 tests
1000 tests
Description
- CoV Protease Substrate-1 EDANS allows for efficient detection of coronavirus (CoV) proteases
- This fluorescence resonance energy transfer (FRET)-based peptide substrate can be cleaved by 3C-like protease (3CLpro), the CoV main protease involved in regulating the coronavirus replication complex
- 3CLpro is also a potential target for therapeutic applications
- CoV Protease Substrate-1 EDANS has an excitation wavelength of 336 nm, an emission wavelength of 455 nm, and an extinction coefficient of 5900 cm-1 M-1
- It consists of a 14 amino acid sequence (KTSAVLQSGFRKME) that also contains the DABCYL (quencher) and EDANS (donor) fragments at the N- and C- termini, respectively
- When the peptide substrate is intact, favorable energetic overlap of the EDANS excited state and the DABCYL absorption leads to efficient intramolecular fluorescence quenching of EDANS by the DABCYL fragment
- Cleavage of the substrate by CoV proteases, however, dissociates the EDANS fragment from DABCYL, restoring the fluorescence of the EDANS moiety that is directly correlated to the CoV protease activity
Specifications
Specifications
| À utiliser avec (application) | Enzyme Substrate; FRET |
| Quantité | 100 tests |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction