Learn More
STEMCELL Technologies CoV Protease Substrate-1 TF5
Click to view available options
Quantité:
100 tests
1 000 tests
Conditionnement:
100 tests
1000 tests
Description
- CoV Protease Substrate-1 TF5 allows for efficient detection of coronavirus (CoV) proteases
- This fluorescence resonance energy transfer (FRET)-based peptide substrate is cleaved by 3C-like proteases (3CLpro), the CoV main protease involved in regulating the coronavirus replication complex
- 3CLpro is also a potential target for therapeutic development
- CoV Protease Substrate-1 TF5 has an excitation wavelength of 649 nm, an emission wavelength of 664 nm, and an extinction coefficient of 250,000 cm-1 M-1
- It consists of a 14 amino acid sequence (KTSAVLQSGFRKME) that also contains the Tide Quencher™ 5 (TQ5; quencher) and Tide Fluor™ 5 (TF5; donor) fragments at the N- and C- termini, respectively
- When the peptide substrate is intact, favorable energetic overlap of the TF5 excited state and the TQ5 absorption leads to efficient intramolecular fluorescence quenching of TF5 by the TQ5 fragment
- Cleavage of the substrate by CoV proteases, however, dissociates the TF5 fragment from TQ5, restoring the fluorescence of the TF5 moiety that is directly correlated to the CoV protease activity
Spécification
Spécification
| À utiliser avec (application) | Enzyme Substrate; FRET |
| Quantité | 1 000 tests |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.