Learn More
Macherey-Nagel™ Cyanuric Acid Test, For Swimming Pools
Allows quick and easy determination of cyanuric acid levels in pool water to help maintain stable chlorine levels. Macherey-Nagel™ Cyanuric Acid Test, For Swimming Pools provides reliable results within 30 seconds—just dip, swirl and read.
Marque: Macherey-Nagel™ 90710
Description
Regular monitoring of water quality in swimming pools enables operators to adjust the dosage of water conditioners in good time, so the pool remains nice and clean. Cyanuric Acid is a chlorine stabilizer that helps pools to remain chlorinated in direct sunlight. It forms a chemically weak bond with chlorine and has the ability to absorb UV light, which helps it to protect chlorine from the sun until the chlorine is consumed for oxidation or disinfection of material in the pool.
Spécification
| Cyanuric Acid | |
| Test strip | |
| 0 - 50 - 100 - 150 - 300mg/L Cya | |
| 25 | |
| 0 - 300mg/L Cya | |
| 30 sec. |
| Orange to Pink | |
| Determination of water quality in swimming pools | |
| Box | |
| 1 Each | |
| 6 x 95 mm | |
| Cyanuric Acid test strip |