Learn More
Thermo Scientific Chemicals Cytochrome C, 90%, from horse heart muscle
Code produit. p-3217580 Tous les produits Thermo Scientific Chemicals Produits
Click to view available options
Quantité:
100 mg
1 g
Conditionnement:
100mg
1g
Code produit. 10571581
Marque: Thermo Scientific Chemicals 147531000
Demander un format en vrac ou personnalisé
Cytochrome C, CAS # 9007-43-6, is a component of the electron transport chain in mitochondria, making it an important electron transporter that forms an electron bridge between respirable substrates and oxygen.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Cytochrome C is a key player of the electron transport chain in mitochondria
• It plays an important role in apoptotic pathways
• It also plays a role in cellular oxidation in animals and plants
which make it an effective biological electron carrier. It plays a vital role in cellular oxidation in animals and plants.
Applications
• Cytochrome c shows peroxidase activity by oxidation of various electron donors such as 2,2-azino-bis (3-ethylbenzthiazoline-6-sulphonic acid) (ABTS), 2-keto-4-thiomethyl butyric acid and 4-aminoantipyrine
• It is assumed to be the functional complex utilized in low-level laser therapy (LLLT), which increases the metabolic activity and frees up more energy for the cells to repair the tissue
• It is used as an important mediator in apoptotic pathways
TRUSTED_SUSTAINABILITY
Spécification
Quantité 100 mg
Conditionnement Glass bottle