Learn More
Dibutyldiethyltin, TRC
Code produit. 30012595
Change view
Click to view available options
Quantité:
100 mg
1 g
Conditionnement:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantité unitSize
30012595 100 mg 100mg
30135088 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit. 30012595 Fournisseur Toronto Research Chemicals Code fournisseur TRCD429455100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 20525-62-6
Nom chimique ou matériau Dibutyldiethyltin
Poids de la formule 292.1213
Formule InChI InChI=1S/2C4H9.2C2H5.Sn/c2*1-3-4-2;2*1-2;/h2*1,3-4H2,2H3;2*1H2,2H3;
Nom IUPAC dibutyl(diethyl)stannane
Formule moléculaire C12 H28 Sn
Poids moléculaire (g/mol) 291.06
SMILES CCCC[Sn](CC)(CC)CCCC
Synonyme Dibutyldiethylstannane, Diethyldibutyltin