Learn More
Diheptyltin Dichloride, TRC
Code produit. 30011544
Change view
Click to view available options
Quantité:
100 mg
1 g
Conditionnement:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantité unitSize
30011544 100 mg 100mg
30138077 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit. 30011544 Fournisseur Toronto Research Chemicals Code fournisseur TRCD450688100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 74340-12-8
Nom chimique ou matériau Di-n-heptyltin-dichloride
Poids de la formule 388.0747
Formule InChI InChI=1S/2C7H15.2ClH.Sn/c2*1-3-5-7-6-4-2;;;/h2*1,3-7H2,2H3;2*1H;/q;;;;+2/p-2
Nom IUPAC dichloro(diheptyl)stannane
Formule moléculaire C14 H30 Cl2 Sn
Poids moléculaire (g/mol) 388
SMILES CCCCCCC[Sn](Cl)(Cl)CCCCCCC
Synonyme Dichlorodiheptylstannane, Diheptyltin dichloride, Di-n-heptyltin dichloride