Learn More
Fisherbrand™ Disposable Inoculating Loops
Tous les produits Fisherbrand Produits
Click to view available options
Volume:
1 μl
10 μl
Stérilité:
Stérile
Quantité:
Carton de 125 (5 paquets de 25/emballage individuel)
Carton de 250 (5 paquets de 50/emballés individuellement)
Conditionnement:
125 pièces
250 pièces
Description
- Ideal for anaerobic microbiology applications where deep agar inoculation is required
- Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
- High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
- Rigid enough to penetrate the media for subsurface inoculation
- Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
- Can be used under hoods without danger
- Color-coded for ease of size identification
Spécification
Spécification
| Jetable | Single-use |
| Volume | 10 μl |
| Matériau | Polystyrène |
| Stérilité | Stérile |
| Quantité | Carton de 125 (5 paquets de 25/emballage individuel) |
| Autoclavable | Not Autoclavable |