Learn More
Invitrogen™ Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 755
Code produit 13427247
Modifier l'affichage
Click to view available options
Quantité:
500 μg
Conditionnement:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
13427247 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 13427247 Fournisseur Invitrogen™ Code fournisseur SA510091
Suivez le workflow : Explorez les économies
Utilisez le code promo 27967
Afficher toutes les promotions
 Article restreint
En stock
Ajouter au panier
Edge
Ajouter au panier
Donkey Polyclonal Secondary Antibody
This antibody is cross-adsorbed and exhibits minimum reactivity to chicken, human, mouse, pig, rabbit and rat.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Goat IgG (H+L) Cross-Adsorbed
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugué DyLight 755
Formule PBS with 0.09% sodium azide; pH 6.8 to 7.4
Espèces hôtes Donkey
Immunogène Goat IgG-heavy and light chain.
Méthode de purification Antigen affinity chromatography
Quantité 500 μg
État réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Goat
Contenu et stockage 4°C
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG