Learn More
Invitrogen™ Donkey anti-Goat IgG (H+L) Secondary Antibody, HRP
Code produit 11595973
Change view
Click to view available options
Quantité:
1 mg
Conditionnement:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11595973 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11595973 Fournisseur Invitrogen™ Code fournisseur PA128664
Donkey Polyclonal Secondary Antibody
Optimal titers should be determined by the researcher.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Goat IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugué HRP
Formule PBS with proprietary stabilizer, 1% BSA and proprietary preservative; pH 7.4
Espèces hôtes Donkey
Immunogène Goat IgG whole molecule.
Méthode de purification Antigen affinity chromatography
Quantité 1 mg
État réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Goat
Contenu et stockage 2-8°C
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG