Learn More
STEMCELL Technologies EasySep™ Human Plasmacytoid DC Isolation Kit
Click to view available options
Quantité:
2 x 109 cellules
Conditionnement:
1 kit
Description
- The EasySep™ Human Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from human peripheral blood mononuclear cells (PBMCs) or leukapheresis by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This product replaces the EasySep™ Human Plasmacytoid DC Enrichment Kit (Catalog #19062), for even faster cell isolations and is compatible with a greater number of EasySep™ magnets
Spécification
Spécification
| Type de cellule | Dendritic Cells |
| À utiliser avec (application) | Cell Isolation |
| Quantité | 2 x 109 cellules |