Learn More
Tocris Bioscience™ Echistatin, alpha1 isoform
GREENER_CHOICE
Code produit. 18016022
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
Code produit. 18016022
Marque: Tocris Bioscience™ 3202/100U
αVβ3 and glycoprotein IIb/IIIa (integrin αIIbβ3) inhibitor
Echistatin, α1 isoform is a potent irreversible αVβ3 integrin antagonist (Ki = 0.27 nM). Disrupts attachment of osteoclasts to bone and inhibits bone reabsorption (IC50 = 0.1 nM). Prevents ADP-induced platelet aggregation via inhibition of glycoprotein IIb/IIIa (GpIIb/IIIa, αIIbβ3) receptors (IC50 = 30 nM) in vitro.
TRUSTED_SUSTAINABILITY
Spécification
Quantité 100 μg