missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EGFR (ErbB1) [d747-749] [A750P], GST Tag, His Tag Recombinant Protein

Code produit. 15980413
Click to view available options
Quantité:
1 mg
10 μg
100 μg
Conditionnement:
100µg
10µg
1mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15980413

Marque: Invitrogen™ A33512

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This EGFR (ErbB1) d747-749 A750P recombinant human protein (672-end) was expressed in insect cells. EGFR (ErbB1) d747-749 A750P recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Cytoplasmic, Mutant.

EGFR receptor tyrosine kinase is a transmembrane glycoprotein. Binding of epidermal growth factor to EGFR induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation, differentiation, motility, and cell survival. Activation of EGFR triggers mitogenic signaling in gastrointestinal mucosa, and its expression is upregulated in colon cancers and most neoplasms. Activation of EGFR triggers activation of the ERK-signaling pathway in normal gastric epithelial and colon cancer cell lines. Inactivation of EGFR with selective inhibitors significantly reduces ERK2 activation, c-fos mRNA expression and cell proliferation. EGFR activation signals multiple downstream signaling cascades such as the Ras - ERK, PI3-K - Akt, Jak - STAT and PKC pathways that help in growth and proliferation of cells. Mutations in the EGFR gene are associated with lung cancer and multiple alternatively spliced transcript variants encode different protein isoforms of EGFR have been found. Increased production or activation of EGFR has been associated with poor prognosis in a variety of tumors. Moreover, EGFR overexpression is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma.
TRUSTED_SUSTAINABILITY

Spécification

Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50mM HEPES with 100mM NaCl, 15mM glutathione, 20% glycerol, 5mM DTT and no preservative; pH 7.5
Poids moléculaire 88.8 kDa
Nom Human EGFR (ErbB1) [d747-749] [A750P], GST Tag, His Tag
Plage de pH 7.5
Méthode de purification Purified
Quantité 10 μg
Conditions de stockage -80°C, Avoid Freeze/Thaw Cycles
État réglementaire RUO
Nom usuel EGFR (d747-749, A750P Mutant)
Famille des kinases EGF Receptors (ErbBs)
Longueur des protéines 672-end
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine GST-tag, His-tag
Séquence 672-end
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Conditions d’expédition Dry Ice
Système d’expression Insect cells
Forme protéique Recombinant, Active, Cytoplasmic, Mutant
Forme Liquid
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis