Learn More
Eptapirone, MedChemExpress
Code produit. 16490438
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
50 mg
Code produit. 16490438
Marque: Medchem Express HY19946
MedChemExpress Eptapirone (F11440) is a potent, selective, high efficacy 5-HT1A receptor agonist with marked anxiolytic and antidepressant potential.
Eptapirone (F11440) is a potent, selective, high efficacy 5-HT1A receptor agonist with marked anxiolytic and antidepressant potential.
Specifications
Nom chimique ou matériau Eptapirone
Forme physique Solid
CAS 179756-58-2
Couleur Blanc
Quantité 5 mg
Notes de qualité de la pureté Research
Formule moléculaire C16H23N7O2
À utiliser avec (application) Neuroscience-Neuromodulation
Synonyme F 11440
Informations sur la solubilité DMSO : 250 mg/mL (723.80 mM; Need ultrasonic)
SMILES O=C(N1C)N(CCCCN2CCN(C3=NC=CC=N3)CC2)N=CC1=O
Poids moléculaire (g/mol) 345.4
Poids de la formule 345.4
Pourcentage de pureté 98.97%
Qualité Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.