Learn More
Tocris Bioscience™ EX 527
GREENER_CHOICE
Code produit. 30001429
Click to view available options
Quantité:
1 mg
10 mg
50 mg
Conditionnement:
10mg
1mg
50mg
Code produit. 30001429
Marque: Tocris Bioscience™ 2780/1
Selective SIRT1 inhibitor
EX 527 is a selective inhibitor of SIRT1 that does not inhibit histone deacetylase (HDAC) or other sirtuin deacetylase family members (IC50 values are 98, 19600, 48700, > 100000 and > 100000 nM for SIRT1, SIRT2, SIRT3, HDAC and NADase respectively). In an animal model of Huntington's disease, EX 527 rescues neuronal degeneration, extends lifespan, reduces huntingtin inclusions and prevents ventricular enlargement. EX 527 enhances p53 acetylation in response to DNA damaging agents. Orally bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 49843-98-3
Formule moléculaire C13H13ClN2O
Spécification
Nom chimique ou matériau EX 527
Quantité 1 mg
Poids de la formule 248.71
Pureté >98%
Poids moléculaire 248.71