Learn More
Applied Biosystems™ Exonuclease I, high concentration, (20 units/μL)
Click to view available options
Quantité:
5 unités
Conditionnement:
1 pièce
Description
Exonuclease I hydrolyzes single-stranded DNA in the 3'→5' direction, releasing. 5'-mononucleotides and leaving the terminal 5'-dinucleotide intact. Hydrolysis is processive and cannot proceed if the 3' terminus is phosphorylated.
Informations sur la commande
Pièce
Spécification
Spécification
| Concentration | 20 U / μl |
| Enzymes | Exonucléase |
| Tampon compatible | Storage Buffer, Glycine Buffer |
| Quantité | 5 unités |
| Type de produit | Exonucléase I |