Learn More
Applied Biosystems™ ExoSAP-IT™ Express PCR Product Clean-up Reagent
ExoSAP-IT™ Express PCR Product Clean-up Reagent removes excess primers and unincorporated nucleotides from a PCR reaction. This enzymatic clean-up method offers precision, increased yield, and a faster turnaround time compared to alternative purification methods, such as spin columns or beads.
Marque: Applied Biosystems™ 75001.40.UL
Description
- Remove unincorporated primers and nucleotides in about 5 minutes
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery, regardless of amplicon length
- Minimize spin column or bead clean-up
For research use only