missing translation for 'onlineSavingsMsg'
Learn More

FCCP, MedChemExpress

Code produit. p-200099359
Encompass_Preferred
Click to view available options
Quantité:
10 mg
50 mg
100 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16482998

Marque: Medchem Express HY100410

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress FCCP is an uncoupler of oxidative phosphorylation (OXPHOS) in mitochondria. FCCP induces activation of PINK1 leading to Parkin Ser65 phosphorylation.

FCCP is an uncoupler of oxidative phosphorylation (OXPHOS) in mitochondria. FCCP induces activation of PINK1 leading to Parkin Ser65 phosphorylation.

Spécification

Nom chimique ou matériau FCCP
CAS 370-86-5
Couleur Jaune
Notes de qualité de la pureté Research
Formule moléculaire C10H5F3N4O
Quantité 10 mg
Synonyme Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone
Informations sur la solubilité DMSO : ≥ 100 mg/mL (393.44 mM) ∣Ethanol : ≥ 33.3 mg/mL (131.01 mM)
SMILES N#C/C(C#N)=N/NC1=CC=C(OC(F)(F)F)C=C1
Poids moléculaire (g/mol) 254.17
Poids de la formule 254.17
Pourcentage de pureté 99.46%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-Kinase/protease
Afficher plus Afficher moins

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.