Learn More
Value Plastics Female Luer to 200 Series Barb
Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,0556 po
0,0625 po
0,125 po
0,188 po
3/32 po
5/32 po
Matériau:
Nylon
Polycarbonate
Polypropylène
Conditionnement:
25 pièces
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Spécification
Spécification
| Certifications/Conformité | Non-DEHP;Non-Phthalate;RoHS |
| Diamètre (impériale) interne | 0,125 po |
| Matériau | Nylon |
| Quantité d’emballage | 25/Pk. |
| Admission | Female Luer x Hose Barb |
Always test the application to verify the performance of the interconnection.