Learn More
Value Plastics Female Luer to 200 Series Barb
Code produit. p-200105067 Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,0556 po
0,0625 po
0,125 po
0,188 po
3/32 po
5/32 po
Matériau:
Nylon
Polycarbonate
Polypropylène
Conditionnement:
25 pièces
Code produit. 30053083
Marque: Value Plastics 5012038
Il en reste null
Ajouter au panier
Ajouter au panier
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Spécification
Certifications/Conformité Non-DEHP;Non-Phthalate;RoHS
Diamètre (impériale) interne 0,125 po
Matériau Nylon
Quantité d’emballage 25/Pk.
Admission Female Luer x Hose Barb
Always test the application to verify the performance of the interconnection.