Learn More
Cole-Parmer Tubulure FEP
Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,0625 po
0,125 po
0,188 po
0,438 po
0,5 po
3/32 po
5/32 po
Diamètre (impériale) externe:
0,25 po
0,5 po
1/8 po
3/16 po
5/32 po
9 à 16 po
Conditionnement:
15m
Description
Made of 100% virgin-grade fluorinated ethylene propylene (FEP). Excellent chemical and thermal resistance. Great optical clarity—easily view your sample in use. Sterilize by autoclave, EtO, and chemical sterilants. Complies with FDA 21 CFR 177.1550, USP Class VI, NSF 51, NSF 61, NSF 372, and ASTM D2116. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Spécification
Spécification
| Certifications/Conformité | ADCF;FDA;NSF-51;NSF-61;RoHS;USP |
| Diamètre (impériale) interne | 0,5 po |
| Diamètre (impériale) externe | 9 à 16 po |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Correction du contenu d'un produit
Nom du produit
Spot an opportunity for improvement?Share a Content Correction