missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB FITC-inulin Green Fluorophore
Click to view available options
Quantité:
100 mg
1 g
5 g
Conditionnement:
100mg
1g
5g
Description
- Product Overview: FITC-inulin is a green-fluorescent, water-soluble polysaccharide tracer derived from plant inulin. It is widely used for quantitative assessment of glomerular filtration rate (GFR), vascular permeability, and diffusion studies. With excitation at 495 nm and emission at 519 nm, FITC-inulin provides stable fluorescence for reliable detection in biological systems without significant cellular uptake or metabolism.
- Features:
- Bright Green Fluorescence: Excitation maximum at 495 nm, emission maximum at 519 nm, suitable for standard fluorescence detection platforms.
- Non-Metabolizable: Inulin is biologically inert, making it ideal for tracking molecular movement without interference from cellular processing.
- High Water Solubility: Fully dissolves in aqueous systems for easy preparation and immediate use in experiments.
- Low Cytotoxicity: Suitable for live-animal, tissue, and cell culture studies without adverse biological effects.
- Reliable Batch-to-Batch Quality: Produced under tight quality control to ensure consistent labeling and fluorescence output.
- Benefits:
- Gold Standard for GFR Measurement: Used in preclinical and experimental models to assess kidney filtration function accurately.
- Reliable Permeability Probe: Ideal for studying vascular barrier function and macromolecular transport in biological tissues.
- Minimal Biological Interference: Inert nature reduces variability and improves experimental reproducibility.
- Stable Fluorescence for Imaging and Quantification: Provides strong, durable fluorescence signals across experimental time courses.
- Applications:
- Glomerular Filtration Rate (GFR) Studies: Quantitative analysis of renal filtration efficiency in animal models.
- Vascular and Tissue Permeability Studies: Tracks movement across biological barriers in health and disease models.
- Tissue Distribution and Diffusion Research: Visualizes the spread of macromolecules in engineered and native tissues.
- Perfusion and Microfluidic Systems: Used in dynamic flow models to monitor molecular transport behavior.
- Technical Specifications:
- Fluorescence: Excitation maximum at 495 nm, emission maximum at 519 nm.
- Appearance: Light-sensitive, greenish dry powder, readily soluble in water.
- Storage and Stability:
- Store dry, protected from light, at 2°C to 8°C to maintain stability and performance.
- Stable under recommended conditions for extended use and reliable fluorescence output.
- Packaging and Availability:
- Supplied as dry powder suitable for long-term storage and ease of handling.
- Available in multiple research-grade pack sizes to meet varying experimental needs.
Spécification
Spécification
| Méthode de détection | Fluorescence |
| Format | Vial |
| Type de produit | Reagent |
| Quantité | 5 g |
| Forme | Crystalline Powder |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu