Learn More
Tocris Bioscience™ Givinostat hydrochloride
Click to view available options
Quantité:
5 mg
25 mg
Conditionnement:
25mg
5mg
Description
Givinostat hydrochloride is a histone deacetylase (HDAC) inhibitor (IC50 = 7.5 to 16 nM). At low concentrations reduces release of pro-inflammatory cytokines, including TNFα, INFγ, IL-1α and IL-1β, from LPS stimulated PBMCs in vitro. Exhibits anti-inflammatory effects in vivo. Improves functional recovery and attenuates tissue damage in a mouse model of traumatic brain injury.
Spécification
Spécification
| Nom chimique ou matériau | [6-[(Diethylamino)methyl]-2-naphthalenyl]methyl-N-[4-[(hydroxyamino)carbonyl]phenyl]carbamate hydrochloride |
| CAS | 199657-29-9 |
| Plage de pourcentage du dosage | >98% |
| Quantité | 25 mg |
| Formule moléculaire | C24H27N3O4.HCl |
| Poids de la formule | Observed MW: 457.95 |