Learn More
STEMCELL Technologies GloCell™ Fixable Viability Dye Red 780
Click to view available options
Quantité:
100 tests
5 x 100 tests
Conditionnement:
100 tests
5 pièces
Description
- GloCell™ Fixable Viability Dye Red 780 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
- The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation
- Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay
- GloCell™ Fixable Viability Dye Red 780 is excited by the red laser at 633 nm and it has a fluorescence emission maximum of 780 nm that can be detected using the 780/60 band pass filter used to detect APC-Cy7
Spécification
Spécification
| À utiliser avec (application) | Flow Cytometry; Immunofluorescence |
| Quantité | 100 tests |