missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TRIM7. The GNIP Recombinant Protein Antigen is derived from E. coli. The GNIP Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-89751. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Specifications
Specifications
| Gene ID (Entrez) | 81786 |
| Espèces | Human |
| Méthode de purification | Chromatography |
| Pureté | >80% |
| Concentration | 0.5mg/mL |
| Contenu et stockage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formule | PBS and 1M Urea, pH 7.4. |
| À utiliser avec (application) | Blocking/Neutralizing, Control |
| Symbole de gène(s) | TRIM7 |
| Type d’étiquette | Unlabeled |
| Show More |
Usage exclusivement réservé à la recherche
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?