Learn More
Invitrogen™ Goat anti-Rabbit IgA Secondary Antibody, HRP
Code produit 11574230
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
11574230 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11574230 Fournisseur Invitrogen™ Code fournisseur PA174362
Please to purchase this item. Need a web account? Register with us today!
Goat Polyclonal Secondary Antibody
PA1-74362 detects Rabbit IgA from rabbit samples and shows no cross-reactivity with other rabbit IG classes in immunoelectrophoresis. Goat anti Rabbit IgA antibody recognizes rabbit IgA and shows no cross-reactivity with other rabbit immunoglobulin classes in immunoelectrophoresis.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Rabbit IgA
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugué HRP
Formule PBS with 0.2% BSA and 0.05% Pro-Clean 400
Espèces hôtes Goat
Immunogène Purified rabbit IgA.
Méthode de purification Affinity chromatography
Quantité 1 mg
État réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Rabbit
Contenu et stockage 4°C, do not freeze
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG