Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, Biotin
Code produit 11543350
Modifier l'affichage
Click to view available options
Quantité:
1.5 mg
Conditionnement:
1.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11543350 1.5 mg 1.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11543350 Fournisseur Invitrogen™ Code fournisseur 31820
Please to purchase this item. Need a web account? Register with us today!
Goat Polyclonal Secondary Antibody
Product # 31820 has been successfully used in Western blot, IF, ICC, ISH, and IHC applications. Product # 31820 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins. This antibody cross-reacts with rabbit IgM (25%). However, this antibody may cross-react with immunoglobulins from other species.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Rabbit IgG (H+L)
Applications ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugué Biotin
Formule PBS with 3mg/mL BSA and 0.08% sodium azide; pH 7.8
Espèces hôtes Goat
Méthode de purification Affinity chromatography
Quantité 1.5 mg
État réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Rabbit
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG