Learn More
Invitrogen™ Human GRK5, GST Tag Recombinant Protein
Code produit. 15448523
Click to view available options
Quantité:
1 mg
100 μg
Conditionnement:
100µg
1mg
Code produit. 15448523
Marque: Invitrogen™ PR7128B
Recombinant Protein
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
GRK5 is a serine/threonine kinase that regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes.
TRUSTED_SUSTAINABILITY
Spécification
Numéro d’adhésion P34947
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Identification génétique (Entrez) 11476, 2869
Poids moléculaire 95.2 kDa
Nom Human GRK5, GST Tag
Quantité 100 μg
État réglementaire RUO - research use only
Alias de gène 1200003L19Rik; Alpha-CP4; AlphaCP-4; FP2025; G protein-coupled receptor kinase 5; G protein-coupled receptor kinase GRK5; Gprk5; GRK; Grk5; GRK5 protein kinase; GRK-pan; Pan-GRK; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4
Nom usuel GRK5
Famille des kinases GRK Kinase Family
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine GST-tag
Séquence Full length
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Système d’expression Insect cells
Forme Liquid