Learn More
Tocris Bioscience™ GSK 626616
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
Description
GSK 626616 is a potent and selective DYRK inhibitor (IC50 = 0.7 nM for DYRK3, similar potency for DYRK1 and 2). Exhibits ∽20-fold selectivity over casein kinase 2 and negligible activity against a panel of 451 kinases screened. Enhances number of CFU-E stimulated by Epo from human marrow. Increases hemoglobin levels in anemic mice. Orally bioavailable.
Spécification
Spécification
| Nom chimique ou matériau | (5Z)-2-[(2,6-Dichlorophenyl)amino]-5-(6-quinoxalinylmethylene)-4(5H)-thiazolone |
| CAS | 1025821-33-3 |
| Quantité | 10 mg |
| Formule moléculaire | C18H10Cl2N4OS |
| Poids de la formule | 401.27 |
| Pureté | 98% |