Learn More
Copper, Red Brass, Gun Metal, ARMI∣MBH
Code produit. 30040364
Click to view available options
Conditionnement:
1 pièce
Code produit. 30040364
Marque: ARMI/MBH MBH33X GM21 B
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Copper, Red Brass, Gun Metal, Ø40x15mm
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Silicon/Phosphorus/Aluminium/Selenium/Nickel/Cadmium/Arsenic/Bismuth/Iron/Silver/Antimony/Tin/Zinc/Lead/Copper
CAS 7440-50-8
Forme Disc
Concentration or Composition (by Analyte or Components) Silicon 0.0213 %/Phosphorus 0.0697 %/Aluminium 0.173 %/Selenium 0.173 %/Nickel 0.197 %/Cadmium 0.249 %/Arsenic 0.333 %/Bismuth 0.459 %/Iron 0.693 %/Silver 0.701 %/Antimony 1.033 %/Tin 4.5 %/Zinc 4.96 %/Lead 7.53 %/Copper 78.86 %