missing translation for 'onlineSavingsMsg'
Learn More

Heme Oxygenase-1-IN-1, MedChemExpress

Code produit. 16440837
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16440837

Marque: Medchem Express HY111798

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress Heme Oxygenase-1-IN-1 (Compound 2) is a heme oxygenase 1 (HO-1) inhibitor with an IC50 of 0.25 μM.

Heme Oxygenase-1-IN-1 (Compound 2) is a heme oxygenase 1 (HO-1) inhibitor with an IC50 of 0.25 μM.

Specifications

Nom chimique ou matériau Heme Oxygenase-1-IN-1
CAS 1093058-52-6
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C13H15BrN2
Quantité 50 mg
Informations sur la solubilité DMSO : 125 mg/mL (447.74 mM; Need ultrasonic)
SMILES BrC1=CC=C(CCCCN2C=CN=C2)C=C1
Poids moléculaire (g/mol) 279.18
Poids de la formule 279.18
Pourcentage de pureté 98.37%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.