missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HEXA Recombinant Protein

Code produit. 16106071
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16106071

Marque: Abnova™ H00003073P01.10ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Human HEXA full-length ORF recombinant protein with GST-tag at N-terminal

This gene encodes the alpha subunit of the lysosomal enzyme β-hexosaminidase that, together with the cofactor GM2 activator protein, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. β-hexosaminidase is composed of two subunits, α and β, which are encoded by separate genes. Both β-hexosaminidase α and β subunits are members of family 20 of glycosyl hydrolases. Mutations in the α or β subunit genes lead to an accumulation of GM2 ganglioside in neurons and neurodegenerative disorders termed the GM2 gangliosidoses. α subunit gene mutations lead to Tay-Sachs disease (GM2-gangliosidosis type I).

  • Theoretical MW (kDa): 83.93
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MTSSRLWFSLLLAAAFAGRATALWPWPQNFQTSDQRYVLYPNNFQFQYDVSSAAQPGCSVLDEAFQRYRDLLFGSGSWPRPYLTGKRHTLEKNVLVVSVVTPGCNQLPTLESVENYTLTINDDQCLLLSETVWGALRGLETFSQLVWKSAEGTFFINKTEIEDFPRFPHRGLLLDTSRHYLPLSSILDTLDVMAYNKLNVFHWHLVDDPSFPYESFTFPELMRKGSYNPVTHIYTAQDVKEVIEYARLRGIRVLAEFDTPGHTLSWGPGIPGLLTPCYSGSEPSGTFGPVNPSLNNTYEFMSTFFLEVSSVFPDFYLHLGGDEVDFTCWKSNPEIQDFMRKKGFGEDFKQLESFYIQTLLDIVSSYGKGYVVWQEVFDNKVKIQPDTIIQVWREDIPVNYMKELELVTKAGFRALLSAPWYLNRISYGPDWKDFYVVEPLAFEGTPEQKALVIGGEACMWGEYVDNTNLVPRLWPRAGAVAERLWSNKLTSDLTFAYERLSHFRCELLRRGVQAQPLNVGFCEQEFEQT

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Spezifikation

Numéro d’adhésion AAH18927
À utiliser avec (application) Antibody Production, Protein Array, ELISA, Western Blot
Formule 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Identification génétique (Entrez) 3073
Poids moléculaire 83.93
Nom HEXA (Human) Recombinant Protein (P01)
Plage de pH 8
Méthode de préparation In vitro wheat germ expression system
Méthode de purification Glutathione Sepharose 4 Fast Flow
Test du contrôle qualité 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantité 10 μg
Source Wheat Germ (in vitro)
Immunogène MTSSRLWFSLLLAAAFAGRATALWPWPQNFQTSDQRYVLYPNNFQFQYDVSSAAQPGCSVLDEAFQRYRDLLFGSGSWPRPYLTGKRHTLEKNVLVVSVVTPGCNQLPTLESVENYTLTINDDQCLLLSETVWGALRGLETFSQLVWKSAEGTFFINKTEIEDFPRFPHRGLLLDTSRHYLPLSSILDTLDVMAYNKLNVFHWHLVDDPSFPYESFTFPELMRKGSYNPVTHIYTAQDVKEVIEYARLRGIRVLAEFDTPGHTLSWGPGIPGLLTPCYSGSEPSGTFGPVNPSLNNTYEFMSTFFLEVSSVFPDFYLHLGGDEVDFTCWKSNPEIQDFMRKKGFGEDFKQLESFYIQTLLDIVSSYGKGYVVWQEVFDNKVKIQPDTIIQVWREDIPVNYMKELELVTKAGFRALLSAPWYLNRISYGPDWKDFYVVEPLAFEGTPEQKALVIGGEACMWGEYVDNTNLVPRLWPRAGAVAERLWSNKLTSDLTFAYERLSHFRCELLRRGVQAQPLNVGFCEQEFEQT
Conditions de stockage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Alias de gène MGC99608/TSD
Nom usuel HEXA
Symbole de gène(s) HEXA
Réactivité croisée Human
Espèces Wheat Germ (in vitro)
Recombinant Recombinant
Marqueur de protéine GST
Forme Solution
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt