Learn More
BOLA HT Replacement Inner Parts
Click to view available options
Diamètre du filetage:
32
45
Diamètre (métrique):
3 mm
3,2 mm
5 mm
6 mm
8 mm
9,5 mm
9,52 mm
12,7 mm
14 mm
16 mm
18 mm
18,01 mm
19,05 mm
20 mm
21 mm
22 mm
30 mm
32 mm
Conditionnement:
1 pièce
Description
Consisting of a v-ring (PPS), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm (not exposed to the medium). Good chemical resistance, the medium is only exposed to PTFE.
Spécification
Spécification
| Certifications/Conformité | FDA |
| Diamètre (métrique) | 5 mm |
| Autoclavable | Autoclavable |
| Diamètre du filetage | 32 |
| Couleur | White |
| Flexibility | Rigid |