missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ADCK4 (aa 180-240) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55566 (PA5-55566. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a protein with two copies of a domain found in protein kinases. The encoded protein has a complete protein kinase catalytic domain, and a truncated domain that contains only the active and binding sites of the protein kinase domain, however, it is not known whether the protein has any kinase activity. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification
Spécification
| Numéro d’adhésion | Q96D53 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 79934 |
| Nom | Human ADCK4 (aa 180-240) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | 0610012P18Rik; aarF domain containing kinase 4; aarF domain-containing protein kinase 4; ADCK4; Atypical kinase COQ8B, mitochondrial; Coenzyme Q protein 8 B; coenzyme Q8B; COQ8B; FLJ12229; NPHS9 |
| Nom usuel | ADCK4 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu