Learn More
Gibco™ Human ANG-1 Recombinant Protein, PeproTech®
Description
130-06-1MG will be provided as 10 x 100 μg (130-06-100UG). 130-06-500UG will be provided as 5 x 100 μg (130-06-100UG). Recombinant Human ANG-1, derived from HeLa cells, is a C-terminal histidine tagged glycoprotein which migrates with an apparent molecular mass of 60.0-70.0 kDa by SDS-PAGE under reducing conditions. Sequencing analysis shows N-terminal sequences starting with Ser-20 and with Asp-70 of the 498 amino acid precursor protein. The calculated molecular weight of Recombinant Human ANG-1 is 56.3 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Spécification
Spécification
| Numéro d’adhésion | Q15389 |
| À utiliser avec (application) | Control, Bioactivity |
| Formule | Protein with no preservative |
| Identification génétique (Entrez) | 284 |
| Poids moléculaire | 56.3 kDa |
| Nom | Human ANG-1 |
| Méthode de purification | Purified |
| Quantité | 100 μg |
| Source | HeLa cells |
| Conditions de stockage | -20°C |
| Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.