missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BAFF (BLyS), Animal-Free Recombinant Protein, PeproTech®

Code produit. 17826658
Click to view available options
Quantité:
250 μg
5 μg
Conditionnement:
250µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17826658

Marque: Gibco™ AF31013250UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human soluble BAFF is a 152 amino acid polypeptide (17.0 kDa), which contains the TNF-like portion of the extracellular domain of BAFF. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q9Y275
À utiliser avec (application) Functional Assay
Format Lyophilized
Formule protein with no preservative
Identification génétique (Entrez) 10673
Poids moléculaire 17 kDa
Nom Human BAFF (BLyS), Animal-Free
Méthode de purification HPLC, SDS-PAGE
Quantité 250 μg
État réglementaire RUO
Concentration en endotoxines <0.1 EU/ μg
Alias de gène ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13 b; tumor necrosis factor (ligand) superfamily, member 13 b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7 A; tumor necrosis factor ligand superfamily member 13 B; Tumor necrosis factor ligand superfamily member 13 b, membrane form; Tumor necrosis factor ligand superfamily member 13 b, soluble form; tumor necrosis factor superfamily member 13 b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4
Nom usuel BAFF (BLyS)
Symbole de gène(s) TNFSF13B
Activité biologique Determined by a mouse splenocyte survival assay. The expected ED50 for this effect is 0.5-2.0 ug/ml
Type de produit Protein
Conjugué Unconjugated
Recombinant Recombinant
Séquence AVQGPEETVT QDCLQLIADS ETPTIQKGSY TFVPWLLSFK RGSALEEKEN KILVKETGYF FIYGQVLYTD KTYAMGHLIQ RKKVHVFGDE LSLVTLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PRENAQISLD GDVTFFGALK LL
Contenu et stockage -20°C
Activité Determined by a mouse splenocyte survival assay. The expected ED50 for this effect is 0.5-2.0 ug/ml
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 95% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis