missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BMP-13/CDMP-2 Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
250µg
500µg
50µg
Description
Recombinant Human BMP-13/CDMP-2 is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
GDF6 is a growth differentiation factor (GDFs), which are members of the transforming growth factor (TGF) superfamily that is involved in embryonic development and adult tissue homeostasis. Both GDF6 and GDF7 are closely related to GDF5 which has been shown to induce activation of plasminogen activator, thereby inducing angiogenesis. It is predominantly expressed in long bones during fetal embryonic development and is involved in bone formation. In Xenopus, GDF6 is expressed at the edge of the neural plate and within the anterior neural plate including the eye fields. GDF6 is required for normal formation of some bones and joints in the limbs, skull, and axial skeleton. It may regulate patterning of the ectoderm by interacting with bone morphogenetic proteins (BMPs), and control eye development. Mutations in this gene result in colobomata, which are congenital abnormalities in ocular development, and in Klippel-Feil syndrome (KFS), a congenital disorder of spinal segmentation.
Spécification
Spécification
| Numéro d’adhésion | Q6KF10 |
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 392255 |
| Poids moléculaire | 27 kDa |
| Nom | Human BMP-13/CDMP-2 |
| Quantité | 250 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu