missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL20 (MIP-3 alpha) Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
Description
Recombinant Human MIP-3alpha is an 8.0 kDa protein containing 70 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
Spécification
Spécification
| Numéro d’adhésion | P78556 |
| À utiliser avec (application) | ELISA, Functional Assay, Western Blot |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 6364 |
| Poids moléculaire | 8 kDa |
| Nom | Human CCL20 (MIP-3 alpha) |
| Quantité | 20 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu