missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL25 (TECK) Recombinant Protein, PeproTech®

Code produit. p-7245912
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17892953

Marque: Gibco™ 30045100UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human TECK is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Chemokines are likely to play an important role in regulating the trafficking of developing T cells within the thymus. Chemokine C-C thymus expressed chemokine (TECK), also designated chemokine ligand 25 (CCL25), small induc-ible cytokine A25, chemokine b-15 or CK b-15, is expressed predominantly in thymic dendritic cells, thymic epithelial cells and in the small intestine. TECK, a CCR9 ligand, has suppressive activity against immature subsets of myeloid progenitors which have been stimulated to proliferate by multiple growth factors. TECK delivers signals through CCR9, which is important for the nav-igation of developing thymocytes. Bone marrow pre-pro-B cells and cells capable of generating pro-B colonies in the presence of interleukin-7 and Flt-3 ligand migrate to TECK, a response lost in later stages of B cell development.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion O15444
À utiliser avec (application) Control, Western Blot Control, ELISA Standard, Binding Assay, Bioactivity
Formule Protein with no preservative
Identification génétique (Entrez) 6370
Poids moléculaire 14.2 kDa
Nom Human CCL25 (TECK)
Méthode de purification Purified
Quantité 100 μg
Source E. coli
Conditions de stockage -20°C
État réglementaire RUO
Concentration en endotoxines <1 EU/μg
Alias de gène A130072A22Rik; AI852536; C-C motif chemokine 25; C-C motif chemokine ligand 25; CCL25; chemokine (C-C motif) ligand 25; chemokine TECK; Ck beta-15; CKb15; Scya25; SCYA25TECKvar; small inducible chemokine 25; small inducible cytokine A25; small inducible cytokine subfamily A (Cys-Cys), member 25; small-inducible cytokine A25; Teck; TECKMGC150327; TECKvar; thymus expressed chemokine; thymus-expressed chemokine
Nom usuel TECK
Symbole de gène(s) CCL25
Activité biologique Determined by its ability to bind CCR9 receptor expressed on the surface of MOLT4 cells.
Type de produit Protein
Conjugué Unconjugated
Recombinant Recombinant
Séquence QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN MQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSKRNVSLL ISANSGL
Contenu et stockage -20°C
Activité Determined by its ability to bind CCR9 receptor expressed on the surface of MOLT4 cells.
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis