missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL8 (MCP-2), Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantité:
2 μg
250 μg
50 μg
Conditionnement:
250µg
2µg
50µg
Description
Recombinant Human MCP-2 (CCL8) is an 8.9 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
Spécification
Spécification
| Numéro d’adhésion | P80075 |
| À utiliser avec (application) | Functional Assay |
| Format | Lyophilized |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 6355 |
| Poids moléculaire | 8.9 kDa |
| Nom | Human CCL8 (MCP-2), Animal-Free |
| Méthode de purification | HPLC, SDS-PAGE |
| Quantité | 250 μg |
| État réglementaire | RUO |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu